missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JAK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
£161.00 - £350.00
Specifications
| Antigen | JAK3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17968782
|
Invitrogen™
PA5145379 |
200 μL |
£350.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17948692
|
Invitrogen™
PA5145380 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Janus kinase 3 (JAK3) phosphorylates members of the STAT (signal transduction and activators of transcription) family. JAK3 is predominantly expressed in immune cells and transduces a signal in response to activation of interleukin receptors. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.Specifications
| JAK3 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| Q62137, Q63272 | |
| 16453, 25326 | |
| Recombinant Mouse JAK3 protein, Leu517-Ala749 (Accession #Q62137). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Breast-JAK3; fae; JAK3; JAK-3; JAK3_HUMAN; JAK3B; JAKL; Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; L JAK; Leukocyte janus kinase; LJAK; L-JAK; RATJAK3; Tyrosine-protein kinase JAK3; wil | |
| JAK3 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| JAK3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title