missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAK3-IN-1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
JAK3-IN-1 is a potent, selective and orally active JAK3 inhibitor with an IC50 of 4.8 nM. JAK3-IN-1 shows over 180-fold more selective for JAK3 than JAK1 (IC50 of 896 nM) and JAK2 (IC50 of 1050 nM).
Specifications
Specifications
| Chemical Name or Material | JAK3-IN-1 |
| CAS | 1805787-93-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H30ClN7O2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 100 mg/mL (196.84 mM; Need ultrasonic) |
| SMILES | O=C(C=C)NC1=CC(CNC2=NC(NC3=C(OC)C=C(N4CCN(C)CC4)C=C3)=NC=C2Cl)=CC=C1 |
| Molecular Weight (g/mol) | 508.02 |
| Formula Weight | 508.02 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction