missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jak2, p Tyr1007, p Tyr1008 Antibody (1A4), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £431.00
Specifications
| Antigen | Jak2 |
|---|---|
| Clone | 1A4 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30032351
|
Novus Biologicals
NBP3-26415-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30023602
|
Novus Biologicals
NBP3-26415-100ul |
100 μL |
£431.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Jak2 Monoclonal antibody specifically detects Jak2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
| Jak2 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2 | |
| A synthesized peptide derived from Human JAK2 [p Tyr1007, p Tyr1008] [UniProt O60674] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1A4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3717 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title