missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jak2 Antibody (53B7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59451JF549
This item is not returnable.
View return policy
Description
Jak2 Monoclonal antibody specifically detects Jak2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| Jak2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2 | |
| Recombinant human JAK2 (1014-1132aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication, Signal Transduction | |
| 3717 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 53B7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction