missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAK1 Mouse anti-Human, Rat, Clone: 3H7A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66466-1-IG-150UL
This item is not returnable.
View return policy
Description
JAK1 (Janus kinase 1) belongs to the family of non-receptor Janus tyrosine kinases, which regulate a spectrum of cellular functions downstream of activated cytokine receptors in the lympho-hematopoietic system. Immunological stimuli, such as interferons and cytokines, induce recruitment of Stat transcription factors to cytokine receptor-associated JAK1. JAK1 then phosphorylates proximal Stat factors, which subsequently dimerize, translocate to the nucleus and bind to cis elements upstream of target gene promoters to regulate transcription. Upon ligand binding, JAK1 undergoes tyrosine phosphorylation and catalytic activation in an interdependent manner. Phosphorylation of tyrosine residues at positions 1022 and 1023 is believed to function in the activation of catalytic events.Specifications
| JAK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P23458 | |
| JAK1 | |
| JAK1 Fusion Protein Ag17940 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3H7A8 | |
| Unconjugated | |
| JAK1 | |
| JAK 1, JAK1, JAK1A, JAK1B, Janus kinase 1, JTK3, Tyrosine protein kinase JAK1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3716, 84598 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction