missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Jagged1 Monoclonal Antibody (C5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551022
This item is not returnable.
View return policy
Description
The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.
Specifications
| Jagged1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P78504, Q63722 | |
| JAG1 | |
| Recombinant Human JAG1 protein, Gly33-Asp250 (Accession #P78504). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 λ |
| Western Blot | |
| C5 | |
| Unconjugated | |
| JAG1 | |
| ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 182, 29146 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction