missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jagged 2 Rabbit anti-Human, Mouse, Clone: 3K1I8, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £395.00
Specifications
| Antigen | Jagged 2 |
|---|---|
| Clone | 3K1I8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351442
|
Bio-Techne
NBP3-15726-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18350853
|
Bio-Techne
NBP3-15726-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Jagged 2 Monoclonal antibody specifically detects Jagged 2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| Jagged 2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Stem Cell Signaling Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 3714 | |
| IgG | |
| Affinity purified |
| 3K1I8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| HJ2, jagged 2, Jagged2, protein jagged-2, SER2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Jagged 2 (Q9Y219). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title