Learn More
Invitrogen™ Jagged 2 Monoclonal Antibody (HMJ2-1), eFluor™ 660, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
£223.00
Specifications
| Antigen | Jagged 2 |
|---|---|
| Clone | HMJ2-1 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
This HMJ2-1 MAb reacts with mouse Jagged-2, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g., Notch 1-4), Jagged-2 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Jagged-2 is expressed by thymic lymphoid and stromal cells, as well as macrophages and dendritic cells in the spleen. Note: Staining of endogenous splenic dendritic (CD8+CD11c+) cells could only be consistently confirmed in cells isolated from C57BL/6, but not Balb/c, mice. Other strains have not been tested. The antiobdy has been reported to crossreact to rat and have blocking function.
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. JAG2 is one of several ligands that activate Notch and related receptors.Specifications
| Jagged 2 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P97607, Q9QYE5 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| HMJ2-1 | |
| Flow Cytometry | |
| eFluor 660 | |
| Armenian Hamster | |
| Mouse, Rat | |
| 16450, 29147 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Jag2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.