Learn More
Jagged 2 Monoclonal Antibody (HMJ2-1), eFluor™ 660, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£79.50 - £208.00
Specifications
Antigen | Jagged 2 |
---|---|
Clone | HMJ2-1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15519936
|
Affymetrix eBioscience
50-3392-82 |
100 μg |
£208.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15509936
|
Affymetrix eBioscience
50-3392-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This HMJ2-1 monoclonal antibody reacts with mouse Jagged-2, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g., Notch 1-4), Jagged-2 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Jagged-2 is expressed by thymic lymphoid and stromal cells, as well as macrophages and dendritic cells in the spleen. Note: Staining of endogenous splenic dendritic (CD8+CD11c+) cells could only be consistently confirmed in cells isolated from C57BL/6, but not Balb/c, mice. Other strains have not been tested. The antiobdy has been reported to crossreact to rat and have blocking function. Applications Reported: This HMJ2-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HMJ2-1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647.
Jagged 2 Monoclonal antibody specifically detects Jagged 2 in Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Jagged 2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
16450, 29147 | |
IgG | |
Affinity chromatography | |
Antibody |
HMJ2-1 | |
Flow Cytometry | |
eFluor 660 | |
Armenian Hamster | |
Mouse, Rat | |
Jag2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Jag2 |
For Research Use Only.