missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jagged 2 Antibody (746513), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4748APCCY7
This item is not returnable.
View return policy
Description
Jagged 2 Monoclonal antibody specifically detects Jagged 2 in Mouse samples. It is validated for Flow Cytometry
Specifications
| Jagged 2 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 746513 | |
| Flow Cytometry | |
| HJ2, jagged 2, Jagged2, protein jagged-2, SER2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Jagged 2, Met27-Leu1084, Accession # Q9QYE5 | |
| 0.1 mL | |
| Cell Cycle and Replication, Stem Cell Signaling Pathway | |
| 3714 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction