missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jagged 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-1161-0.25MG
This item is not returnable.
View return policy
Description
Jagged 1 Polyclonal specifically detects Jagged 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Jagged 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
| AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, jagged 1, Jagged1, JAGL1, protein jagged-1 | |
| Rabbit | |
| 0.25 mg | |
| 182 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:20000-1:100000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P78504 | |
| JAG1 | |
| This protein A purified Jagged 1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids near the N-terminus of human Jagged-1 protein. | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction