missing translation for 'onlineSavingsMsg'
Learn More

Jagged 1 Antibody (J1G53-3) - Azide and BSA Free, Novus Biologicals™

Product Code. 18372263 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372263 100 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372263 Supplier Novus Biologicals Supplier No. NBP311671

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Jagged 1 Monoclonal specifically detects Jagged 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Surface Plasmon Resonance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Jagged 1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone J1G53-3
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Surface Plasmon Resonance
Formulation 0.2 um-filtered solution in PBS, pH 7.4
Gene Alias AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, jagged 1, Jagged1, JAGL1, protein jagged-1
Host Species Mouse
Immunogen Recombinant human Jagged 1.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Growth and Development, Neuronal Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 182
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.