missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jagged 1 Antibody (188331) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1277JF635
This item is not returnable.
View return policy
Description
Jagged 1 Monoclonal antibody specifically detects Jagged 1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Jagged 1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, jagged 1, Jagged1, JAGL1, protein jagged-1 | |
| NS0-derived recombinant human Jagged 1, Ser32-Asp296, Accession # P78504 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Growth and Development, Neuronal Cell Markers, Stem Cell Signaling Pathway | |
| 182 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 188331 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction