missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ Jackson Safety™ G40 Polyurethane Coated Gloves

Product Code. 15353342
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
60 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15353342 Large 60 pairs
15343342 Medium 60 pairs
15363342 X-Large 60 pairs
15373342 2X-Large 60 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15353342 Supplier KleenGuard™ Supplier No. 13839

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Code Black
Color Black
Material Polyurethane
Product Type Gloves
Size Large
For Use With (Application) Ideally suited use in precision assembly, electronics assembly, controls handling, packaging, and inspection
Quantity 60Pair
Resistant To Abrasion and tear
Series G40
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.