missing translation for 'onlineSavingsMsg'
Learn More

Jaceosidin, MedChemExpress

Product Code. 16469118
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469118 10 mg 10mg
16412559 1 mg 1mg
16403916 25 mg 25mg
16401528 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469118 Supplier Medchem Express Supplier No. HYN0831
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Jaceosidin is a flavonoid isolated from Artemisia vestita, induces apoptosis in cancer cells, activates Bax and down-regulates Mcl-1 and c-FLIP expression. Jaceosidin exhibits anti-cancer, anti-inflammatory activities, decreases leves of inflammatory markers, and suppresses COX-2 expression and NF-κB activation.

Jaceosidin is a flavonoid isolated from Artemisia vestita, induces apoptosis in cancer cells, activates Bax and down-regulates Mcl-1 and c-FLIP expression. Jaceosidin exhibits anti-cancer, anti-inflammatory activities, decreases leves of inflammatory markers, and suppresses COX-2 expression and NF-κB activation.

Specifications

Chemical Name or Material Jaceosidin
CAS 18085-97-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H14O7
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (378.46 mM; Need ultrasonic) ∣Ethanol : 7.14 mg/mL (21.62 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(O)C(OC)=C2)OC3=CC(O)=C(OC)C(O)=C13
Molecular Weight (g/mol) 330.29
Formula Weight 330.29
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.