missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAB1 Rabbit anti-Human, Mouse, Rat, Clone: 3X5F0, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £395.00
Specifications
| Antigen | JAB1 |
|---|---|
| Clone | 3X5F0 |
| Dilution | Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18353844
|
Novus Biologicals
NBP3-16328-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18310892
|
Novus Biologicals
NBP3-16328-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JAB1 Monoclonal antibody specifically detects JAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| JAB1 | |
| Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis), COP9 signalosome complex subunit 5, CSN5COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 5, EC 3.4, JAB138 kDa Mov34 homolog, Jun activation domain-binding protein 1, MOV-34, SGN5MGC3149, Signalosome subunit 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human JAB1 (Q92905). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3X5F0 | |
| ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10987 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title