Learn More
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint
Product Code. 15814764
Click to view available options
Type:
Male Spherical Joint
Joint Size:
12/2
12/5
18/7
18/9
28/12
28/15
35/20
40/25
51/35
Unit Size:
Each
Product Code. 15814764
Brand: GPE Scientific™ GBM/18/9
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
  • Spherical joint designed for use with cup joint
  • Available with different joint sizes, shank bores, and shank ODs
Specifications
Item Description Shank Bore: 9 mm, Shank OD: 12 mm
Type Male Spherical Joint
Joint Size 18/9