Learn More
GPE Scientific™ J Young O-Rings for Spherical Joints
Click to view available options
Size:
12/2 and 12/5 spherical joints
18/7 and 18/9 spherical joints
28/12 and 28/15 spherical joints
35/20 size spherical joints
40/25 size spherical joints
51/35 size spherical joints
Material:
PTFE-Covered Viton
Viton
Unit Size:
Each
Description
- Suitable for use with spherical joints
- Available in Viton or PTFE-coated Viton
- Available in different joint sizes
Specifications
Specifications
| Product Type | O-ring for Spherical Joint |
| Material | Viton |
| Size | 18/7 and 18/9 spherical joints |