missing translation for 'onlineSavingsMsg'
Learn More

J.T. Baker™ BAKER ANALYZED™ Eosin Y (yellowish)

Product Code. 15558784 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15558784 25 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558784 Supplier JT Baker Supplier No. L08803

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Certified Stain for histology

  • For Laboratory, Research, or Manufacturing Use
  • Management of Change (MOC) category = NI
  • Certified for histology and compounding of blood stains. Lot analysis on label.

Specifications

Absorbance 1.0 mg/200 mL in 0.01% Na2CO3,1-cm path
Melting Point 295°C to 296°C
CAS 17372-87-1
Bio. Stain Comm. Certification Certified by the Biological Stain Commission
Molecular Formula C20H6Br4Na2O5
Linear Formula C20H6Br4Na2O5
MDL Number MFCD00005040
Synonym Eosin yellowish , Tetrabromofluorescein disodium salt , Eosin Y disodium salt , Eosin 225 , Solvent Red 43 , Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate , 2,4,5,7-Tetrabromofluorescein disodium salt , Field's Stain B
InChI Key SEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
IUPAC Name disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Weight (g/mol) 691.86
Packaging Wide Mouth Amber Glass Bottle, Black T/E Cap
Quantity 25 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.