Learn More
YMC Group J'Sphere ODS-M80, Classical Analytical HPLC Column (4.6 mm ID), 8 nm, S-4 µm, 250 x 4.6 mm
Brand:  YMC Group JM08S04-2546WT
Encompass_Preferred
J'Sphere ODS-M80, Classical Analytical HPLC Column (4.6 mm ID), 8 nm, S-4 µm, 250 x 4.6 mm
Product Code. 14201333
  • £1243.08 / Each