missing translation for 'onlineSavingsMsg'
Learn More

J-147, MedChemExpress

Product Code. p-200102008
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478507

Brand: Medchem Express HY13779

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress J-147 is an exceptionally potent, orally active, neuroprotective agent for cognitive enhancement. J-147 can readily pass the blood brain barrier (BBB). J-147 can inhibit monoamine oxidase B (MAO B) and the dopamine transporter with EC50 values of 1.88 μM and 0.649 μM, respectively. J-147 has potential for the treatment of Alzheimer’s disease (AD).

J-147 is an exceptionally potent, orally active, neuroprotective agent for cognitive enhancement. J-147 can readily pass the blood brain barrier (BBB). J-147 can inhibit monoamine oxidase B (MAO B) and the dopamine transporter with EC50 values of 1.88 μM and 0.649 μM, respectively. J-147 has potential for the treatment of Alzheimer’s disease (AD).

Specifications

Chemical Name or Material J-147
CAS 1146963-51-0
Color Pink
Purity Grade Notes Research
Molecular Formula C18H17F3N2O2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (285.44 mM)
SMILES O=C(N(C1=CC=C(C)C=C1C)/N=C/C2=CC=CC(OC)=C2)C(F)(F)F
Molecular Weight (g/mol) 350.34
Formula Weight 350.34
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.