missing translation for 'onlineSavingsMsg'
Learn More

J-147, MedChemExpress

Product Code. 16478507
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478507 5 mg 5mg
16422007 50 mg 50mg
16415858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478507 Supplier Medchem Express Supplier No. HY13779

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress J-147 is an exceptionally potent, orally active, neuroprotective agent for cognitive enhancement. J-147 can readily pass the blood brain barrier (BBB). J-147 can inhibit monoamine oxidase B (MAO B) and the dopamine transporter with EC50 values of 1.88 μM and 0.649 μM, respectively. J-147 has potential for the treatment of Alzheimer’s disease (AD).

J-147 is an exceptionally potent, orally active, neuroprotective agent for cognitive enhancement. J-147 can readily pass the blood brain barrier (BBB). J-147 can inhibit monoamine oxidase B (MAO B) and the dopamine transporter with EC50 values of 1.88 μM and 0.649 μM, respectively. J-147 has potential for the treatment of Alzheimer’s disease (AD).

Specifications

Chemical Name or Material J-147
CAS 1146963-51-0
Color Pink
Purity Grade Notes Research
Molecular Formula C18H17F3N2O2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (285.44 mM)
SMILES O=C(N(C1=CC=C(C)C=C1C)/N=C/C2=CC=CC(OC)=C2)C(F)(F)F
Molecular Weight (g/mol) 350.34
Formula Weight 350.34
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.