missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IZUMO4 Polyclonal specifically detects IZUMO4 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | IZUMO4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | C19orf36, chromosome 19 open reading frame 36, IMAGE:4215339, IZUMO family member 4, izumo sperm-egg fusion protein 4, putative SAMP14 binding protein variant 1, putative SAMP14 binding protein variant 2, Sperm 22 kDa protein c113 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of IZUMO4 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?