missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IZUMO4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17130804 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17130804 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17130804 Supplier Invitrogen™ Supplier No. PA5112942

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IZUMO4 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

IZUMO4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IZUMO4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IZUMO4
Gene Accession No. Q1ZYL8
Gene Alias 9030607L17Rik; AI550407; C19orf36; IMAGE:4215339; IZUMO family member 4; izumo sperm-egg fusion protein 4; Izumo4; putative SAMP14 binding protein variant 1; putative SAMP14 binding protein variant 2; RGD1564722; sperm 22 kDa protein c113; UNQ831/PRO1758
Gene Symbols IZUMO4
Host Species Rabbit
Immunogen Recombinant Human Izumo sperm-egg fusion protein 4 protein (16-214AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113177, 362832
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.