missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IZUMO1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£225.00 - £435.00
Specifications
| Antigen | IZUMO1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunocytochemistry |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18458430
|
Novus Biologicals
NBP1-83086-25ul |
25 μL |
£225.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286178
|
Novus Biologicals
NBP1-83086 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IZUMO1 Polyclonal specifically detects IZUMO1 in Human, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.Specifications
| IZUMO1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 284359 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VVLALKSLEKDYLPGHLDAKHHKAMMERVENAVKDFQELSLNEDAYMGVVDEATLQKGSWSLLKDLKRITDSDVKGDLFVKELFWMLHLQKETFATYVARFQKEAYCPNKCGVMLQTLIWCKNCKKEVHACRKSYDCGERNVEVPQM | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunocytochemistry | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ61440, IZUMO, izumo sperm-egg fusion 1, izumo sperm-egg fusion protein 1, MGC34799, OBF, Oocyte binding/fusion factor, Sperm-specific protein izumo | |
| IZUMO1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title