missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IZUMO1 Polyclonal antibody specifically detects IZUMO1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ61440, IZUMO, izumo sperm-egg fusion 1, izumo sperm-egg fusion protein 1, MGC34799, OBF, Oocyte binding/fusion factor, Sperm-specific protein izumo |
| Immunogen | IZUMO1 antibody was raised against an 18 amino acid peptide near the center of human IZUMO1. The immunogen is located within amino acids 170 - 220 of IZUMO1. Amino Acid Squence: ELNWHQASEGLTDYSFY |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 284359 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?