missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IZUMO1 Polyclonal specifically detects IZUMO1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | IZUMO1 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | FLJ61440, IZUMO, izumo sperm-egg fusion 1, izumo sperm-egg fusion protein 1, MGC34799, OBF, Oocyte binding/fusion factor, Sperm-specific protein izumo |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant IZUMO1 (AAH34769.1; Met1-Arg292). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?