missing translation for 'onlineSavingsMsg'
Learn More

Ixazomib, TRC

Product Code. 30134474
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134474 50 mg 50mg
30137081 100 mg 100mg
30004843 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134474 Supplier Toronto Research Chemicals Supplier No. TRCI94200050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072833-77-2
Molecular Formula C14 H19 B Cl2 N2 O4
Molecular Weight (g/mol) 361.03
Synonym Boronic acid, B-[(1R)-1-[[2-[(2,5-dichlorobenzoyl)amino]acetyl]amino]-3-methylbutyl]-, B-[(1R)-1-[[2-[(2,5-Dichlorobenzoyl)amino]acetyl]amino]-3-methylbutyl]boronic acid, Ixazomib, MLN 2238, Ninlaro
IUPAC Name [(1R)-1-[[2-[(2,5-dichlorobenzoyl)amino]acetyl]amino]-3-methylbutyl]boronic acid
SMILES CC(C)C[C@H](NC(=O)CNC(=O)c1cc(Cl)ccc1Cl)B(O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H19BCl2N2O4/c1-8(2)5-12(15(22)23)19-13(20)7-18-14(21)10-6-9(16)3-4-11(10)17/h3-4,6,8,12,22-23H,5,7H2,1-2H3,(H,18,21)(H,19,20)/t12-/m0/s1
Formula Weight 360.0815
Chemical Name or Material Ixazomib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.