missing translation for 'onlineSavingsMsg'
Learn More

Ixazomib, MedChemExpress

Product Code. 16450337
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16450337 50 mg 50mg
16436697 5 mg 5mg
16412936 25 mg 25mg
16403988 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16450337 Supplier Medchem Express Supplier No. HY10453
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ixazomib (MLN2238) is a selective, potent, and reversible proteasome inhibitor, which inhibits the chymotrypsin-like proteolytic (β5) site of the 20S proteasome with an IC50 of 3.4 nM (Ki of 0.93 nM).

Ixazomib (MLN2238) is a selective, potent, and reversible proteasome inhibitor, which inhibits the chymotrypsin-like proteolytic (β5) site of the 20S proteasome with an IC50 of 3.4 nM (Ki of 0.93 nM).

Specifications

Chemical Name or Material Ixazomib
Physical Form Solid
CAS 1072833-77-2
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H19BCl2N2O4
For Use With (Application) Cancer-Kinase/protease
Synonym MLN2238
Solubility Information DMSO : 62.5 mg/mL (173.12 mM; Need ultrasonic)
SMILES OB([C@@H](NC(CNC(C1=CC(Cl)=CC=C1Cl)=O)=O)CC(C)C)O
Molecular Weight (g/mol) 361.03
Formula Weight 361.03
Percent Purity 96.67%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.