missing translation for 'onlineSavingsMsg'
Learn More

Ixabepilone, MedChemExpress

Product Code. 16430107
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430107

Brand: Medchem Express HY10222

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Ixabepilone (BMS-247550) is an orally bioavailable microtubule inhibitor, which binds to tubulin and promotes tubulin polymerization and microtubule stabilization, thereby arrests cells in the G2-M phase of the cell cycle and induces tumor cell apoptosis.

Ixabepilone (BMS-247550) is an orally bioavailable microtubule inhibitor, which binds to tubulin and promotes tubulin polymerization and microtubule stabilization, thereby arrests cells in the G2-M phase of the cell cycle and induces tumor cell apoptosis.

Specifications

Chemical Name or Material Ixabepilone
Physical Form Solid
CAS 219989-84-1
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C27H42N2O5S
For Use With (Application) Cancer-programmed cell death
Synonym BMS-247550 Aza-epothilone B
Solubility Information DMSO : 83.33 mg/mL (164.46 mM; Need ultrasonic)
SMILES O=C([C@@H]([C@H]([C@H](CCC[C@]1(O[C@]1(C[C@H](N2)/C(C)=C/C3=CSC(C)=N3)[H])C)C)O)C)C(C)([C@H](CC2=O)O)C
Molecular Weight (g/mol) 506.7
Formula Weight 506.7
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.