missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ixabepilone, MedChemExpress
Description
Ixabepilone (BMS-247550) is an orally bioavailable microtubule inhibitor, which binds to tubulin and promotes tubulin polymerization and microtubule stabilization, thereby arrests cells in the G2-M phase of the cell cycle and induces tumor cell apoptosis.
Specifications
Specifications
| Chemical Name or Material | Ixabepilone |
| Physical Form | Solid |
| CAS | 219989-84-1 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C27H42N2O5S |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | BMS-247550 Aza-epothilone B |
| Solubility Information | DMSO : 83.33 mg/mL (164.46 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?