missing translation for 'onlineSavingsMsg'
Learn More

IWS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen IWS1
Concentration 0.25 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16836734
View Documents
Proteintech
16943-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16826734
View Documents
Proteintech
16943-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.
Specifications

Specifications

IWS1
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q3SWT4, Q8C1D8, Q96ST2
291705, 55677, 73473
IWS1 Fusion Protein Ag10554
Primary
-20°C
IWS1
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
IWS1, IWS1 homolog (S. cerevisiae), IWS1 like protein, IWS1L, Protein IWS1 homolog
Iws1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.