missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IWS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557374
This item is not returnable.
View return policy
Description
Immunogen sequence: IFGESGDEEE EEFTGFNQED LEEEKGETQV KEAEDSDSDD NIKRGKHMDF LSDFEMMLQR KKSMSGKRRR NRDGGTFISD ADDVVSAMIV KMNEAA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.
Specifications
| IWS1 | |
| Polyclonal | |
| Unconjugated | |
| IWS1 | |
| 1700069O15Rik; 3000008H23; AL117927; AW214353; interacts with Spt6; IWS1; IWS1 homolog; IWS1 homolog (S. cerevisiae); IWS1, SUPT6H interacting protein; Iws1l; IWS1-like protein; Protein IWS1 homolog; RGD1304762 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55677 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96ST2 | |
| IWS1 | |
| Recombinant protein corresponding to Human IWS1. Recombinant protein control fragment (Product #RP-96178). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction