missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies IWR-1-endo
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
- IWR-1-endo potently inhibits WNT signaling by blocking a cell-based WNT/β-catenin pathway reporter response with an IC50 value of 180 nM (Chen et al.) It inhibits WNT-induced accumulation of β-catenin, through stabilization of the destruction complex member AXIN2 (Chen et al.)
- Promotes self-renewal and maintains pluripotency of human embryonic stem cells and mouse Epi-stem cells when used in combination with CHIR99021 (Kim et al.) DIFFERENTIATION
- Promotes differentiation of cardiomyocytes from human pluripotent stem cells (PSCs) that have been induced to mesoderm by addition of Activin A and/or BMP4 (Ren et al.; Willems et al.)
- Induces the differentiation of human PSC-derived alveolar epithelial type II (AETII) to AETI cells (Ghaedi et al.)
Specifications
Specifications
| CAS | 1127442-82-3 |
| Quantity | 5 mg |
| Molecular Formula | C25H19N3O3 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction