missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IWP L6
GREENER_CHOICE

Product Code. 18738817
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18738817

Brand: Tocris Bioscience™ 4992/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent PORCN inhibitor

IWP L6 is a potent inhibitor of Porcupine (PORCN), a membrane-bound O-acyltransferase (MBOAT) (EC50 = 0.5 nM). Shown to inhibit Wnt-dependent processes such as tailfin regeneration and posterior axis formation in zebrafish; also reduces branching morphogenesis in cultured mouse embryonic kidneys.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type IWP L6
Purity >98%
Molecular Weight (g/mol) 472.58
Inhibitors IWP L6

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.