missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ IWP 2

Product Code. 18083912
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18083912 10 mg 10mg
18035402 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18083912 Supplier R&D Systems™ Supplier No. 3533/10
  • ONLINE EXCLUSIVE PRICE
    £225.18
    £200.55 / 10mg
    Save £24.63
    10% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PORCN inhibitor; inhibits Wnt processing and secretion

IWP 2 is a potent inhibitor of Wnt processing and secretion (IC50 = 27nM). IWP 2 inactivates PORCN, a membrane-bound O-acyltransferase (MBOAT), and selectively inhibits palmitoylation of Wnt. Blocks Wnt-dependent phosphorylation of Lrp6 receptor and Dvl2, and β-catenin accumulation. IWP 2 suppresses self-renewal in R1 embryonic stem cells and promotes cardiomyocyte differentiation from hPSCs.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 686770-61-6
Molecular Formula C22H18N4O2S3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 466.6
Purity >98%
Molecular Weight (g/mol) 466.59
Chemical Name or Material IWP 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.