missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies IWP-2
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- IWP-2 inhibits the WNT pathway (IC50 = 27 nM) at the level of the pathway activator Porcupine
- Porcupine is a membrane-bound acyltransferase that palmitoylates WNT proteins, which leads to WNT secretion and signaling capability
- (Chen et al.), Willert et al.)
- Suppresses self-renewal of mouse embryonic stem (ES) cells and supports their conversion to epiblast-like stem cells (ten Berge et al.)
- Inhibits maintenance and proliferation of mouse Lgr5+ intestinal and cochlear epithelial stem cells, demonstrating the importance of WNT signaling in these processes (Chai et al.), Farin et al.)
- Promotes cardiomyocyte differentiation from human pluripotent stem cells (Lian et al.), Minami et al.)
Specifications
Specifications
| CAS | 686770-61-6 |
| Synonym | Inhibitor of WNT Production-2 |
| Quantity | 1 mg |
| Molecular Formula | C22H18N4O2S3 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction