missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ivosidenib

Description
Ivosidenib is a potent inhibitor of mutant isocitrate dehydrogenase enzyme 1 (mIDH1), with activity against a variety of mutant IDH1 enzymes (IC50 range = 5-19 nM). Reduces intracellular 2-HG (Cat. No. 6122) by 96% at 0.5 μM. Increases sensitivity of melanoma to Temozolomide (Cat. No. 2706) in vivo and inhibits wild-type IDH1 in low magnesium and low nutrient levels. Ivosidenib also inhibits growth of tumors in cell and animal models of pancreatic cancer and inhibits invasion and migration of chondrosarcoma cells in vitro.
Specifications
Specifications
| Chemical Name or Material | (2S)-N-{(1S)-1-(2-Chlorophenyl)-2-[(3,3-difluorocyclobutyl)amino]-2-oxoethyl}-1(4-cyanopyridin-2-yl)-N-(5-fluoropyridin-3-yl)-5-oxopyrrolidine-2-carboxamide |
| CAS | 1448347-49-6 |
| Quantity | 10 mg |
| Target | Isocitrate Dehydrogenase 1/IDH1 |
| Molecular Formula | C28H22ClF3N6O3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?