missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IVNS1ABP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14741-1-AP-150UL
This item is not returnable.
View return policy
Description
IVNS1ABP is a protein coding gene involved in many cell functions, including pre-mRNA splicing, the aryl hydrocarbon receptor (AHR) pathway, F-actin organization and protein ubiquitination. Plays a role in the dynamic organization of the actin skeleton as a stabilizer of actin filaments by association with F-actin through Kelch repeats. Protects cells from cell death induced by actin destabilization. Functions as modifier of the AHR/Aryl hydrocarbon receptor pathway increasing the concentration of AHR available to activate transcription. In addition, functions as a negative regulator of BCR(KLHL20) E3 ubiquitin ligase complex to prevent ubiquitin-mediated proteolysis of PML and DAPK1, two tumor suppressors. Inhibits pre-mRNA splicing (in vitro).; (Microbial infection) involved in the alternative splicing of influenza A virus M1 mRNA through interaction with HNRNPK, thereby facilitating the generation of viral M2 protein.Specifications
| IVNS1ABP | |
| Polyclonal | |
| Unconjugated | |
| IVNS1ABP | |
| ARA3, FLARA3, HSPC068, IVNS1ABP, KIAA0850, ND1, NS 1, NS1, NS1 binding protein, NS1 BP, NS1BP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10625, 117198, 289089 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q920Q8, Q9Y6Y0 | |
| Ivns1abp | |
| IVNS1ABP Fusion Protein Ag6412 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction