missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IVNS1ABP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551926
This item is not returnable.
View return policy
Description
Immunogen sequence: QDELIEKPMS PMQYARSGLG TAEMNGKLIA AGGYNREECL RTVECYNPHT DHWSFLAPMR TPRARFQMAV LMGQLYVVGG SNGHSDDLSC GEMYDSNIDD WIPVPELRTN RCNAGVCALN GKLYIVGGSD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
IVNS1ABP is a protein coding gene involved in many cell functions, including pre-mRNA splicing, the aryl hydrocarbon receptor (AHR) pathway, F-actin organization and protein ubiquitination. Plays a role in the dynamic organization of the actin skeleton as a stabilizer of actin filaments by association with F-actin through Kelch repeats. Protects cells from cell death induced by actin destabilization. Functions as modifier of the AHR/Aryl hydrocarbon receptor pathway increasing the concentration of AHR available to activate transcription. In addition, functions as a negative regulator of BCR(KLHL20) E3 ubiquitin ligase complex to prevent ubiquitin-mediated proteolysis of PML and DAPK1, two tumor suppressors. Inhibits pre-mRNA splicing (in vitro).; (Microbial infection) involved in the alternative splicing of influenza A virus M1 mRNA through interaction with HNRNPK, thereby facilitating the generation of viral M2 protein.
Specifications
| IVNS1ABP | |
| Polyclonal | |
| Unconjugated | |
| IVNS1ABP | |
| 1190004M08Rik; 1700126I16Rik; AA960440; ARA3; aryl hydrocarbon receptor-associated 3; aryl hydrocarbon receptor-associated protein 3; FLARA3; HSPC068; influenza virus NS1A binding protein; influenza virus NS1A-binding protein; Influenza virus NS1A-binding protein homolog; Ivns1abp; kelch family protein Nd1; kelch family protein Nd1-L; kelch-like family member 39; Kelch-like protein 39; KIAA0850; KLHL39; mKIAA0850; NCX downstream gene 1; ND1; Nd1L; Nd1-L; ND1-L2; Nd1S; Nd1-S; NS1; NS-1; NS1-binding protein; NS1-binding protein homolog; Ns1bp; NS1-BP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6Y0 | |
| IVNS1ABP | |
| Recombinant protein corresponding to Human IVNS1ABP. Recombinant protein control fragment (Product #RP-102250). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction