missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IVNS1ABP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100582
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IVNS1ABP.
IVNS1ABP is a protein coding gene involved in many cell functions, including pre-mRNA splicing, the aryl hydrocarbon receptor (AHR) pathway, F-actin organization and protein ubiquitination. Plays a role in the dynamic organization of the actin skeleton as a stabilizer of actin filaments by association with F-actin through Kelch repeats. Protects cells from cell death induced by actin destabilization. Functions as modifier of the AHR/Aryl hydrocarbon receptor pathway increasing the concentration of AHR available to activate transcription. In addition, functions as a negative regulator of BCR(KLHL20) E3 ubiquitin ligase complex to prevent ubiquitin-mediated proteolysis of PML and DAPK1, two tumor suppressors. Inhibits pre-mRNA splicing (in vitro).; (Microbial infection) involved in the alternative splicing of influenza A virus M1 mRNA through interaction with HNRNPK, thereby facilitating the generation of viral M2 protein.
Specifications
| IVNS1ABP | |
| Polyclonal | |
| Unconjugated | |
| IVNS1ABP | |
| 1190004M08Rik; 1700126I16Rik; AA960440; ARA3; aryl hydrocarbon receptor-associated 3; aryl hydrocarbon receptor-associated protein 3; FLARA3; HSPC068; influenza virus NS1A binding protein; influenza virus NS1A-binding protein; Influenza virus NS1A-binding protein homolog; Ivns1abp; kelch family protein Nd1; kelch family protein Nd1-L; kelch-like family member 39; Kelch-like protein 39; KIAA0850; KLHL39; mKIAA0850; NCX downstream gene 1; ND1; Nd1L; Nd1-L; ND1-L2; Nd1S; Nd1-S; NS1; NS-1; NS1-binding protein; NS1-binding protein homolog; Ns1bp; NS1-BP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10625, 117198, 289089 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q920Q8, Q9Y6Y0 | |
| IVNS1ABP | |
| A synthesized peptide derived from human IVNS1ABP(Accession Q9Y6Y0), corresponding to amino acid residues T403-R453. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction