missing translation for 'onlineSavingsMsg'
Learn More

Ivermectin, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18752474
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18752474

Brand: Tocris Bioscience 1260/100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allosteric modulator of α7 nicotinic receptors

Positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor and the purinergic P2X4 receptor. Antihelmintic and antiviral. Also modulates glutamate- and GABA-activated chloride channels. Potentiates glycine-gated currents at low concentrations (30 nM). Inhibits replication of SARS-CoV-2 in vitro (IC50 ∽ 2μM).

This product is composed of both Ivermectin B1a (>90%) and Ivermectin B1b.

TRUSTED_SUSTAINABILITY

Specifications

CAS 70288-86-7
Molecular Formula C48H74O14
Quantity 100 mg
Synonym ivermectin
InChI Key AZSNMRSAGSSBNP-RDKIRRGNSA-N
SMILES CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C
Molecular Weight (g/mol) 875.106
PubChem CID 122172888
Formula Weight 875.1
Percent Purity >96%
Chemical Name or Material Ivermectin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.