missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IVD Mouse anti-Human, Clone: 2D12A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66032-1-IG-20UL
This item is not returnable.
View return policy
Description
Isovaleryl-CoA dehydrogenase (IVD) is a mitochondrial matrix enzyme that catalyzes the third step in leucine catabolism. The genetic deficiency of IVD results in an accumulation of isovaleric acid, which is toxic to the central nervous system and leads to isovaleric acidemia.Specifications
| IVD | |
| Monoclonal | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26440 | |
| IVD | |
| IVD Fusion Protein Ag18042 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2D12A5 | |
| Unconjugated | |
| IVD | |
| ACAD2, IVD | |
| Mouse | |
| Protein A | |
| RUO | |
| 3712 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction