missing translation for 'onlineSavingsMsg'
Learn More

IVD Antibody (OTI1B10), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18414474 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414474 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414474 Supplier Novus Biologicals Supplier No. NBP271055AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IVD Monoclonal antibody specifically detects IVD in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IVD
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1B10
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial
Host Species Mouse
Immunogen Full length recombinant protein of human IVD (NP_002612) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3712
Target Species Human, Mouse, Rat, Canine, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.