missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IVD Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18798G
This item is not returnable.
View return policy
Description
IVD Polyclonal antibody specifically detects IVD in Human samples. It is validated for ELISA
Specifications
| IVD | |
| Polyclonal | |
| ELISA | |
| ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial | |
| Produced in rabbits immunized with purified recombinant Human IVD (His30-His423, Accession: AAH17202.1) | |
| 0.1 mL | |
| Stem Cell Markers | |
| 3712 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction