missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IVD Antibody (04), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06536MFV450
This item is not returnable.
View return policy
Description
IVD Monoclonal antibody specifically detects IVD in Human samples. It is validated for Flow Cytometry
Specifications
| IVD | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 4 | |
| Flow Cytometry | |
| ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IVD (AAH17202.1; His30-His423). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 3712 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction