missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ivacaftor, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Ivacaftor (VX-770) is a potent and orally bioavailable CFTR potentiator, targeting G551D-CFTR and F508del-CFTR with EC50s of 100 nM and 25 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Ivacaftor |
| CAS | 873054-44-5 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H28N2O3 |
| Quantity | 5 mg |
| Synonym | VX-770 |
| Solubility Information | DMSO : 50 mg/mL (127.39 mM; Need ultrasonic) |
| SMILES | CC(C)(C1=C(C=C(C(C(C)(C)C)=C1)O)NC(C2=CNC3=C(C2=O)C=CC=C3)=O)C |
| Molecular Weight (g/mol) | 392.49 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction