missing translation for 'onlineSavingsMsg'
Learn More

Ivacaftor, MedChemExpress

Product Code. 16468197
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16468197

Brand: Medchem Express HY13017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ivacaftor (VX-770) is a potent and orally bioavailable CFTR potentiator, targeting G551D-CFTR and F508del-CFTR with EC50s of 100 nM and 25 nM, respectively.

Ivacaftor (VX-770) is a potent and orally bioavailable CFTR potentiator, targeting G551D-CFTR and F508del-CFTR with EC50s of 100 nM and 25 nM, respectively.

Specifications

Chemical Name or Material Ivacaftor
CAS 873054-44-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H28N2O3
Quantity 5 mg
Synonym VX-770
Solubility Information DMSO : 50 mg/mL (127.39 mM; Need ultrasonic)
SMILES CC(C)(C1=C(C=C(C(C(C)(C)C)=C1)O)NC(C2=CNC3=C(C2=O)C=CC=C3)=O)C
Molecular Weight (g/mol) 392.49
Formula Weight 392.49
Percent Purity 99.08%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.