missing translation for 'onlineSavingsMsg'
Learn More

Ivacaftor Carboxylic Acid, TRC

Product Code. 30012132
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012132 1 mg 1mg
30010487 10 mg 10mg
30011616 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012132 Supplier Toronto Research Chemicals Supplier No. TRCI9406051MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246213-24-0
Molecular Formula C24 H26 N2 O5
Molecular Weight (g/mol) 422.47
Synonym Benzeneacetic acid, 4-[[(1,4-dihydro-4-oxo-3-quinolinyl)carbonyl]amino]-5-(1,1-dimethylethyl)-2-hydroxy-α,α-dimethyl-, 4-[[(1,4-Dihydro-4-oxo-3-quinolinyl)carbonyl]amino]-5-(1,1-dimethylethyl)-2-hydroxy-α,α-dimethylbenzeneacetic acid, Ivacaftor carboxylic acid, Ivacaftor impurity 1, 2-(5-tert-Butyl-2-hydroxy-4-(4-oxo-1,4-dihydroquinoline-3-carboxamido)phenyl)-2-methylpropanoic acid
IUPAC Name 2-[5-tert-butyl-2-hydroxy-4-[(4-oxo-1H-quinoline-3-carbonyl)amino]phenyl]-2-methylpropanoic acid
SMILES CC(C)(C)c1cc(c(O)cc1NC(=O)C2=CNc3ccccc3C2=O)C(C)(C)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C24H26N2O5/c1-23(2,3)15-10-16(24(4,5)22(30)31)19(27)11-18(15)26-21(29)14-12-25-17-9-7-6-8-13(17)20(14)28/h6-12,27H,1-5H3,(H,25,28)(H,26,29)(H,30,31)
Formula Weight 422.1842
Percent Purity >95%
Chemical Name or Material Ivacaftor carboxylic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.