missing translation for 'onlineSavingsMsg'
Learn More

Ivabradine hydrochloride, MedChemExpress

Product Code. 16498136
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498136 100 mg 100mg
16494257 50 mg 50mg
16478368 10 mg 10mg
16461386 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498136 Supplier Medchem Express Supplier No. HYB0162A
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ivabradine hydrochloride is a potent and orally active HCN (hyperpolarization-activated cyclic nucleotide-gated) channel blocker that inhibits the cardiac pacemaker current (If). Ivabradine hydrochloride reduces dose-dependently heart rate without modification of blood pressure. Ivabradine hydrochloride shows anticonvulsant, anti-ischaemic and anti-anginal activity.

Ivabradine hydrochloride is a potent and orally active HCN (hyperpolarization-activated cyclic nucleotide-gated) channel blocker that inhibits the cardiac pacemaker current (If). Ivabradine hydrochloride reduces dose-dependently heart rate without modification of blood pressure. Ivabradine hydrochloride shows anticonvulsant, anti-ischaemic and anti-anginal activity.

Specifications

Chemical Name or Material Ivabradine hydrochloride
CAS 148849-67-6
Color White
Purity Grade Notes Research
Molecular Formula C27H37ClN2O5
Quantity 100 mg
Solubility Information H2O : 50 mg/mL (99.00 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (49.50 mM; Need ultrasonic)
SMILES O=C1N(CCCN(C[C@@H]2C3=CC(OC)=C(OC)C=C3C2)C)CCC4=CC(OC)=C(OC)C=C4C1.Cl
Molecular Weight (g/mol) 505.05
Formula Weight 505.05
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.